Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 17beta-Estradiol Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco E0601ML

Catalog No. 01-001-290


Add to Cart

Description

Description

A Certified Spiking Solution suitable for use as starting material in calibrators, controls, or linearity standards for a variety of LC/MS or GC/MS applications from endocrinology to clinical and diagnostic testing. Estradiol is a sex hormone prescribed under the trade names Vagifem™, Estring™, Evamist, Elestrin™, Estrace™, and Estrogel™ as a treatment for hypoestrogenism.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 50-28-2
MFCD00003693
1.0 mg/mL in acetonitrile
Ampule
2°C (35.6°F) - closed cup
C18H24O2
UN 1648 3 / PGII
272.38
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation