Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 18-Hydroxycorticosterone Solution, 100 μg/mL in Acetonitrile:Water (9:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001455 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-455 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-455 Supplier MilliporeSigma Supelco Supplier No. H1061ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This Certified Spiking Solution™ is suitable for multiple LC/MS applications in clinical and diagnostic testing and endocrinology. 18-Hydroxycorticosterone is tested along with a panel of other related hormones including cortisol, aldosterone, and 11-deoxycortisol in neonates and children for many endocrine diseases from congenital adrenal hyperplasia (CAH) to various hormone enzyme deficiencies of the adrenal glands and steroid/hormone pathways.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 561-65-9
Linear Formula C21H30O5
MDL Number MFCD00042685
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile:water (9:1)
Formula Weight 362.46
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.