Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (1S,2R)-(+)-Ephedrine-d3 Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco E0261ML

Catalog No. 01-001-285


Add to Cart

Description

Description

A stable-labeled internal standard suitable for quantitation of ephedrine levels by LC/MS or GC/MS for a variety of testing applications from clinical toxicology or forensic analysis to quality control testing of dietary supplements containing ephedrine. Ephedrine is a methamphetamine analog commonly used as a stimulant and appetite suppressant. It is also found as an active constituent in herbs used for traditional Chinese medicine, such as má huáng.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 285979-73-9
MFCD01075446
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C10H13ClNOD3
UN 1230 - class 3 - PG 2 - Methanol, solution
204.71
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation