Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 2-Hydroxyethylflurazepam-d4 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001481 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-481 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-481 Supplier MilliporeSigma Supelco Supplier No. H9191ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

2-Hydroxyethylflurazepam is a primary metabolite of the benzodiazepine flurazepam, sold under the trade names Falmane and Salmadorm for treatment of mild to moderate insomnia. This stable-labeled internal standard is suitable for quantitation of hydroxyethylflurazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1397209-35-6
Linear Formula C17D4H10ClFN2O2
MDL Number MFCD01861539
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 336.78
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.