Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 2-Hydroxyethylflurazepam Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001393 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-393 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-393 Supplier MilliporeSigma Supelco Supplier No. F9021ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

2-Hydroxyethylflurazepam is a major urinary metabolite of the benzodiazepine flurazepam. Flurazepam is marketed under the trade names Dalmane and Dalmadorm for treatment of mild to moderate insomnia. This Certified Snap-N-Spike™ Solution is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 20971-53-3
Linear Formula C17H14ClFN2O2
MDL Number MFCD00869360
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 332.76
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.