Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 2-Oxo-3-hydroxy-LSD Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001806 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-806 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-806 Supplier MilliporeSigma Supelco Supplier No. O0131ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

2-Oxo-3-hydroxy-LSD is a major urinary metabolite of lysergic acid diethyl amide, or LSD. This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. Known for its role in 1960's American counterculture, LSD is one of the most potent psychedelic drugs ever created.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 111295-09-1
Linear Formula C20H25N3O3
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Synonym 2-Oxo-3-hydroxy-LSD solution, 2-Oxo-3-hydroxy-lysergic acid diethylamide, 2-Oxo-3-hydroxy-lysergic acid diethylamide solution
Formula Weight 355.43
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.