Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 25B-NBOMe Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001048 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-048 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-048 Supplier MilliporeSigma Supelco Supplier No. B0611ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

25B-NBOMe is a second generation 2C amine derivative of the phenethylamine 2C-B. Sold under street names such as New Nexus, N-Bomb, and 2CB this designer drug is a very potent hallucinogen with active doses in the microgram range. Suitable uses for this certified Snap-N-Spike™ solution include as starting material for calibrators or controls in 25B-NBOMe testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C18H22BrNO3 · HCl
MDL Number MFCD30186957
UN Number UN 1230 - class 3 - PG 2 - Methanol
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 416.74
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.