Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 3,3',5-Triiodo-L-thyronine-13C6 Solution, 100 μg/mL in Methanol with 0.1N NH3, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0771ML

Catalog No. 01-002-074


Add to Cart

Description

Description

A Certified Spiking Solution of the internal standard 3,3′,5-Triiodo-L-thyronine-13C6 (T3-13C6). 3,3′,5-Triiodo-L-thyronine or T3 is a thyroid hormone involved with many physiological processes in the human body including growth and development, metabolism, body temperature and heart rate. This internal standard is suitable for quantitation of free and total T3 serum levels by LC-MS/MS for determination of hyperthyroidism caused by excess T3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1213431-76-5
UN 1230 - class 3 - PG 2 - Methanol, solution
656.93
1 mL
13C6C9H12I3NO4
100 μg/mL in methanol with 0.1N NH3
Ampule
9.7°C (49.5°F)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation