Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 3,3',5'-Triiodo-L-thyronine-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0781ML

Catalog No. 01-002-075


Add to Cart

Description

Description

Reverse T3 or rT3 is a biologically inactive form of the thyroid hormone T3 and is produced in the body particularly during periods of stress. Under certain conditions such as fibromyalgia, liver disease, and chronic fatigue syndrome (CFS), more rT3 is produced which inhibits the desirable conversion of thyroxine (T4) to T3. This internal standard and Certified Spiking Solution™ is suitable for quantitation of reverse T3 serum concentrations by LC-MS/MS to monitor impact of these and other diseases on thyroid hormone levels.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1786403-77-7
UN 1230 - class 3 - PG 2 - Methanol, solution
656.93
1 mL
13C6C9H12I3NO4
100 μg/mL in methanol with 0.1N NH3
Ampule
9.7°C (49.5°F)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation