Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 3,3',5'-Triiodo-L-thyronine Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002072 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-072 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-072 Supplier MilliporeSigma Supelco Supplier No. T0751ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Reverse T3, also known as rT3 or 3,3′,5′-Triiodo-L-thyronine, is a biologically inactive form of the thyroid hormone T3 and is produced in the body particularly during periods of stress. Under certain conditions such as fibromyalgia, liver disease, and chronic fatigue syndrome (CFS), more rT3 is produced which minimizes the desirable conversion of thyroxine (T4) to T3. Reverse T3 serum concentrations are measured by LC-MS/MS to monitor impact of these and other diseases on thyroid hormone levels.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 5817-39-0
Linear Formula C15H12I3NO4
MDL Number MFCD00036864
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol with 0.1N NH3
Synonym 3,3′,5′-Triiodo-L-thyronine, Reverse T3, rT(3)
Formula Weight 650.97
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.