Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 3,3',5-Triiodo-L-thyronine (T3) Solution, 100 μg/mL in Methanol with 0.1N NH3, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002071 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-071 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-071 Supplier MilliporeSigma Supelco Supplier No. T0741ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

T3 or 3,3′,5-Triiodo-L-thyronine is a thyroid hormone involved with many physiological processes in the human body including growth and development, metabolism, body temperature and heart rate. Free and total T3 serum levels are measured by LC-MS/MS for determination of hyperthyroidism caused by excess T3. This Certified Spiking Solution™ is suitable for use as a starting material in preparation of linearity standards, calibrators, and controls for thyroid hormone testing methods by LC-MS/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 6893-02-3
Melting Point 234 to 238°C (lit.)
Linear Formula C15H12I3NO4
MDL Number MFCD00002593
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol with 0.1N NH3
Synonym 3,3′,5-Triiodo-L-thyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, Liothyronine, T3
Formula Weight 650.97
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.