Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 3,4-Methylenedioxypyrovalerone-d8 HCl (MDPV-d8 HCl) Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M1501ML

Catalog No. 01-001-638


Add to Cart

Description

Description

An internal standard of 3,4-Methylenedioxypyrovalerone, a widely known bath salt and designer drug with cases of abuse reported throughout the US and Europe. Also known as MDPV, Mtv, or Super Coke, the illicit drug started appearing in the US in 2010 as a legal drug alternative sold under names including Ivory Wave, Tranquility, and Cloud 9.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1246820-09-6
UN 1230 - class 3 - PG 2 - Methanol, solution
3,4-Methylenedioxypyrovalerone-d8 hydrochloride, MDPV-D8 HCl
Ampule
9.7°C (49.5°F) - closed cup
C16D8H13NO3 · HCl
100 μg/mL in methanol (as free base)
319.85
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation