Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 3'-Hydroxystanozolol-D3, , Sold by MilliporeSigma™ Supelco™

Catalog No. 01001976 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-976 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-976 Supplier MilliporeSigma Supelco Supplier No. S0705MG

missing translation for 'validMainframeMsg'

Certified reference material

3′-Hydroxystanozolol is a major urinary metabolite of stanozolol, an anabolic androgenic steroid. Stanozolol is used as a treatment for osteoporosis and steroid-defficiency but has also been abused in sports as a performance-enhancing drug. This internal standard is suitable for quantitation of 3′-Hydroxystanozolol levels in urine, serum, or plasma by LC/MS or GC/MS for sports testing, urine drug testing, or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 170082-17-4
Linear Formula C21H29D3N2O2
Formula Weight 347.51
Packaging Vial
Quantity 5 mg

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.