Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 3-Trifluoromethylphenylpiperazine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002055 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-055 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-055 Supplier MilliporeSigma Supelco Supplier No. T0451ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Snap-N-Spike™ reference Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research. 3-Trifluoromethylphenylpiperazine, or TFMPP, is a recreational drug and stimulant sold illicitly under the street name “Legal X.” TFMPP is a structural analog of benzylpiperazine or BZP.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 16015-69-3
Linear Formula C11H14ClF3N2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym 3-Trifluoromethylphenylpiperazine hydrochloride, TFMPP
Formula Weight 266.69
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.