Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 4-ANPP-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001038 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-038 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-038 Supplier MilliporeSigma Supelco Supplier No. A1770.5ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This stable-labeled internal standard is suitable for quantification of 4-ANPP levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing applications. 4-ANPP, also known as despropionyl fentanyl, is an intermediate in the synthesis of fentanyl and related opioids and is often found as an impurity in fentanyl preparations.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C1313C6H24N2
Concentration 100 μg/mL in methanol
Formula Weight 286.36
Packaging Ampule
Quantity 0.5 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.