Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 4-Chloromethcathinone (4-CMC HCl) Hydrochloride Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001126 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-126 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-126 Supplier MilliporeSigma Supelco Supplier No. C1741ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

4-Chloromethcathinone, also known as 4-CMC, is a stimulant and designer drug found in 'bath salts' sold at head shops and over the Internet. These synthetic cathinones offer recreational uses that mimic the effects of illegal drugs such as cocaine, methamphetamine, and LSD. This certified solution standard is suitable for numerous GC/MS and LC/MS applications including clinical toxicology testing, forensic analysis, urine drug testing or calibrator preparation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C10H12ClNO · HCl
MDL Number MFCD29036742
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1 mg/mL in methanol (as free base)
Formula Weight 234.12
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.