Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 4-Hydroxy-3-methoxyphenyl-D3-acetic-D2 Acid Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco H0921ML

Catalog No. 01-001-445


Add to Cart

Description

Description

Homovanillic acid (HVA) is a urinary metabolite of the catecholamine dopamine and is associated with dopamine levels in the brain. This stable labeled internal standard is suitable for quantitating HVA levels in LC/MS applications such as clinical and diagnostic testing or endocrinology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 53587-32-9
UN 1230 - class 3 - PG 2 - Methanol, solution
4-Hydroxy-3-methoxyphenyl-d3-acetic-d2 Acid (HVA-d5), HVA-D5 solution
Ampule
9.7°C (49.5°F) - closed cup
C9H5D5O4
100 μg/mL in methanol
187.20
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -70°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation