Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 4-Methyl-N-ethyl-pentedrone (4-MEAP HCl) Hydrochloride Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001672 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-672 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-672 Supplier MilliporeSigma Supelco Supplier No. M1991ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

4-Methyl-N-ethyl-pentedrone, also known as 4-MEAP and 4-MEAPP, is a stimulant drug of the cathinone class and has emerged on the illicit drug market as a designer drug. 4-MEAP has been found to be incorrectly sold as 4-methylpentedrone. This Certified Spiking Solution is suitable as starting material in calibrators and controls for GC/MS or LC/MS testing applications in forensic analysis, urine drug testing or clinical toxicology.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 18297-05-7
Linear Formula C14H21NO · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1 mg/mL in methanol (as free base)
Formula Weight 255.78
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.