Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-4-Methylephedrine-D3 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M1601ML

Catalog No. 01-001-646


Add to Cart

Description

Description

A bath salt cathinone analog and primary urinary metabolite of mephedrone. Mephedrone, also known as 4-methylmethcathinone, is a synthetic cathinone and stimulant widely found in numerous bath salt blends across the US, Europe, and Asia. This certified Snap-N-Spike™ solution is suitable as an internal standard for quantitation of (±)-4-Methylephedrine by LC/MS or GC/MS for applications in urine drug testing, clinical toxicology, or forensic testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
218.74
1 mL
C11H14D3NO · HCl
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation