Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001743 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-743 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-743 Supplier MilliporeSigma Supelco Supplier No. N0761ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Certified Spiking Solution™ suitable for use in nicotine testing, environmental monitoring, or clinical and diagnostic testing of nicotine biomarkers by LC-MS/MS or GC/MS. Also known as nicotine-derived nitrosamine ketone, NNK is a potent carcinogen and biomarker of exposure to cigarette smoke. NNK, produced upon the curing of tobacco, is found in a variety of tobacco products including chewing tobacco, snuff, cigarettes, and cigars.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 64091-91-4
Linear Formula C10H13N3O2
MDL Number MFCD00274580
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym 4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, 4-(Methylnitrosamino)-1-(3-pyridinyl)-1-butanone, NNK
Formula Weight 207.23
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.