Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 5alpha-Dihydrotestosterone (DHT) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0731ML

Catalog No. 01-001-193


Add to Cart

Description

Description

Also known as androstanolone or DHT, 5α-Dihydrotestosterone is an androgen or male sex hormone that has been implicated in a broad range of disease from male pattern baldness to benign prostatic hyperplasia and prostate cancer. As an anabolic steroid, DHT has also been used as a performance-enhancing drug to promote muscle growth. This Certified Spiking Solution™ is suitable for GC/MS and LC/MS applications from clinical and diagnostic testing and endocrinology to sports testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 521-18-6
MFCD00003667
1.0 mg/mL in methanol
290.44
1 mL
C19H30O2
UN 1230 - class 3 - PG 2 - Methanol, solution
5alpha-Dihydrotestosterone solution, 5-alpha-Dihydrotestosterone, DHT, Dihydrotestosterone, 5alpha-DHT solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -70°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation