Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 6alpha-Naloxol Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001761 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-761 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-761 Supplier MilliporeSigma Supelco Supplier No. N1031ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

6α-Naloxol is a metabolite of naloxone, an opiate drug used to counter the effects of opioid overdose. This certified reference solution is suitable as starting material for preparation of calibrators and controls in naloxone testing by LC/MS or GC/MS for forensic analysis, urine drug testing, pharmaceutical research or clinical toxicology applications. In the US, naloxone has become more widely accessible to the general public as a result of state, federal and local initiatives developed to decrease heroin overdose deaths.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 20410-95-1
Linear Formula C19H23NO4
MDL Number MFCD12031398
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 329.39
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.