Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 6beta-Hydroxytestosterone-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002048 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-048 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-048 Supplier MilliporeSigma Supelco Supplier No. T0341ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

6β-Hydroxytestosterone-D3 is an internal standard suitable for quantitation of testosterone levels in urine by LC/MS or GC/MS for applications from clinical and diagnostic testing to endocrinology and sports testing. 6β-Hydroxytestosterone is a major urinary metabolite of testosterone, a hormone from the androgen group. Testosterone is an important sex hormone and anabolic steroid used as a performance enhancing drug.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C19H25O3D3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 307.44
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.