Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 6beta-Hydroxytestosterone Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco H0591ML

Catalog No. 01-001-433


Add to Cart

Description

Description

A Certified Spiking Solution™ suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical and diagnostic testing to endocrinology and sports testing. 6β-Hydroxytestosterone is a major urinary metabolite of testosterone, a hormone from the androgen group of steroids. Testosterone is an important sex hormone and anabolic steroid used as a performance-enhancing drug.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 62-99-7
MFCD00171612
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C19H28O3
UN 1230 - class 3 - PG 2 - Methanol, solution
304.42
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation