Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 6beta-Naltrexol-3-beta-D-glucuronide, 1.0 mg/mL (20% Water in Acetonitrile), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001762 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-762 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-762 Supplier MilliporeSigma Supelco Supplier No. N1041ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

6β-Naltrexol-3-β-D-glucuronide is a primary urinary metabolite of naltrexone, an opiate drug used to counter the effects of opioid overdose. Naltrexone is sold under the trade names such as Revia, Depade and Vivitrol. This Certified Spiking Solution™ is suitable for a variety of LC/MS or GC/MS applications, including forensic toxicology analysis, urine drug testing, and hydrolysis controls.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 1391506-73-2
Linear Formula C26H33NO10
MDL Number MFCD12031424
Concentration 1.0 mg/mL (20% water in acetonitrile)
Formula Weight 519.54
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.