Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 6beta-Naltrexol Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001716 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-716 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-716 Supplier MilliporeSigma Supelco Supplier No. N0381ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

6β-Naltrexol is the primary urinary metabolite of Naltrexone, an opiate sold under the trade names Revia, Depade, and Vivitrol™ and used primarily in the management of alcohol or opiate dependence. This analytical reference standard is appropriate for use in LC/MS or GC/MS applications from clinical toxicology and forensic analysis to urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 49625-89-0
Linear Formula C20H25NO4
MDL Number MFCD00871803
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 343.42
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.