Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 7-Hydroxymitragynine-D3 Solution, 100 μg/mL in Methanol with 0.1N NH3, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001458 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-458 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-458 Supplier MilliporeSigma Supelco Supplier No. H1091ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

An internal standard suitable for quantitation of 7-Hydroxymitragynine levels in MS-based applications including clinical toxicology, urine drug testing or forensic analysis. Along with mitragynine, 7-Hydroxymitragynine is an active alkaloid found in the plant Mitragynine speciosa, commonly known as Kratom. The compound possesses opioid agonistic activity with a potency 30- and 17-fold higher than that of mitragynine and morphine, respectively.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C23H27D3N2O5
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol with 0.1N NH3
Formula Weight 417.51
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.