Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 7-Hydroxyquetiapine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001439 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-439 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-439 Supplier MilliporeSigma Supelco Supplier No. H0811ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Certified Solution Standard for the primary active metabolite of quetiapine, marketed as the atypical antipsychotic Seroquel™ for the treatment of depression, bipolar disorder, and schizophrenia. This new reference standard is suitable for a variety of GCMS and LCMS applications from forensic and forensic toxicology analysis to urine drug testing. We also offer certified Snap-N-Shoot™ solutions of the parent drug Quetiapine fumarate (Q-001) and the internal standard Quetiapine-D8 hemifumarate (Q-002) as well as a wide selection of other atypical antipsychotics and their metabolites and internal standards, such as aripiprazole, clozapine, risperidone, olanzapine, and ziprasidone.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 139079-39-3
Linear Formula C21H25N3O3S
MDL Number MFCD09260019
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 399.51
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.