Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 9-Hydroxyrisperidone-D4 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco H0951ML

Catalog No. 01-001-446


Add to Cart

Description

Description

An internal standard suitable for use in LC/MS or GC/MS applications including forensic analysis, clinical toxicology, urine drug testing, or isotope dilution methods. Also known as paliperidone, 9-Hydroxyrisperidone is sold under the brand name Invega, an atypical antipsychotic used for acute and maintenance treatment of schizophrenia. The drug is also the primary metabolite of risperidone, marketed as the atypical antipsychotic Risperdal™.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1020719-55-4
MFCD09955571
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C23D4H23FN4O3
UN 1230 - class 3 - PG 2 - Methanol, solution
430.51
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation