Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ AB-FUBINACA Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001975 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-975 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-975 Supplier MilliporeSigma Supelco Supplier No. S0651ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

AB-FUBINACA is a potent cannabinoid receptor agonist sold as a synthetic cannabinoid in Spice herbal blends. This certified reference solution is suited for use in synthetic cannabinoid testing methods by LC/MS o GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1185282-01-2
Linear Formula C20H21FN4O2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 368.40
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.