Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ AB-PINACA, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco S0821ML

Catalog No. 01-001-983


Add to Cart

Description

Description

A potent CB1and CB2 receptor agonist, AB-PINACA is a member of the aminoalkylindazole class of synthetic cannabinoids. According to the NFLIS 2014 midyear report, AB-PINACA accounted for 11% of all synthetic cannabinoid drug reports in the United States during the first half of 2014. This certified reference solution is suited for use in synthetic cannabinoid testing methods by LC/MS or GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1445752-09-9
MFCD24368588
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C18H26N4O2
UN 1230 - class 3 - PG 2 - Methanol, solution
330.42
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation