Learn More
CERILLIANT™ Acryl fentanyl-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco A1560.5ML
Catalog No. 01-001-027
Add to Cart
Description
Description
This stable-labeled internal solution standard is suitable for quantitation of acryl fentanyl levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, urine drug testing or forensic analysis applications. Acryl fentanyl, also known as acryloylfentanyl, is a highly potent opioid analgesic fentanyl analog that is sold as designer drug and abused recreationally.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
67-56-1
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C22H21D5N2O · HCl
375.95
0.5 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation