Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ alpha-Hydroxymidazolam-d4 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco H9211ML

Catalog No. 01-001-482


Add to Cart

Description

Description

α-Hydroxymidazolam is an active, major metabolite in urine and blood of the benzodiazepine midazolam. Midazolam is sold as Dormicum, Hypnovel™, and Versed as a sedative and treatment for insomnia and seizures. This internal standard is suitable for midazolam testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, pain prescription monitoring or forensic analysis applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
MFCD08276448
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C18D4H9ClFN3O
UN 1230 - class 3 - PG 2 - Methanol, solution
345.79
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation