Learn More
CERILLIANT™ alpha-Lactalbumin (from bovine milk), certified reference material, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco L045100MG
Catalog No. 01-001-549
Add to Cart
Description
Description
α-Lactalbumin is an important whey protein that regulates the production of lactose in milk and is present in the milk of almost all mammalian species. This full-length protein CRM is suitable as starting material for use in calibrators or controls for a variety of LC-MS/MS testing applications including allergen testing, infant formula testing and dietary or nutritional testing applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
9051-29-0
Ampule
100 mg
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation