Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ alpha-Pyrrolidinovalerophenone-D8 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco P1011ML

Catalog No. 01-001-870


 Restricted Item
Add to Cart

Description

Description

This Certified Spiking Solution™ is suitable as an internal standard for α-PVP testing methods by GC/MS or LC/MS in forensic analysis, clinical toxicology, or urine drug testing applications. Also known as α-Pyrrolidinovalerophenone or O-2387, α-PVP is a derivative of the synthetic cathinone and bath salt MDPV. This stimulant drug has been found along with MDPV in several bath salt blends across the US, Europe, and Australia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
alpha-PVP hydrochloride solution
Ampule
9.7°C (49.5°F) - closed cup
C15H13D8NO · HCl
100 μg/mL in methanol (as free base)
275.84
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation