Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-alpha-Tocopherol (Vitamin E) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002145 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-145 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-145 Supplier MilliporeSigma Supelco Supplier No. V0201ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

α-Tocopherol is a lipid-soluble form of Vitamin E important to human health as an antioxidant. Often the result of poor nutrition or low absorption of dietary fat, vitamin E defficiency is monitored in patient serum samples by LC-MS/MS or HPLC.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 10191-41-0
Linear Formula C29H50O2
MDL Number MFCD00006848
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Density 0.950 g/mL at 20°C (lit.)
Synonym (+/-)-alpha-Tocopherol, DL-all-rac-alpha-Tocopherol, Vitamin E
Formula Weight 430.71
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.