Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Amitriptyline-d3 Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco A1211ML

Catalog No. 01-001-011


Add to Cart

Description

Description

Amitriptyline, marketed under brand names such as Elavil, Tryptizol, or Laroxyl, is a tricyclic antidepressant used in the treatment of major depression and neuropathic pain. This certified Snap-N-Spike™ solution is suitable as an internal standard for quantitation of amitriptyline by LC/MS or GC/MS in clinical toxicology, forensic analysis, urine drug testing, isotope dilution methods, or medication monitoring applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 342611-00-1
UN 1230 - class 3 - PG 2 - Methanol, solution
316.88
1 mL
C20D3H21ClN
1.0 mg/mL in methanol (as free base)
Ampule
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation