Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Aprobarbital Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco A1501ML

Catalog No. 01-001-025


Add to Cart

Description

Description

Aprobarbital, also known as aprobarbitone, is sold as Oramon, Somnifaine and Allonal. Aprobarbital has sedative, hypnotic and anticonvulsant properties and was primarily used for the treatment of insomnia. This Snap-N-Spike Reference Solution is applicable for use in applications including clinical toxicology, medication monitoring, urine drug testing, and forensic analysis by LC-MS/MS or GC/MS.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 77-02-1
MFCD00057559
1 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C10H14N2O3
UN 1230 - class 3 - PG 2 - Methanol, solution
210.23
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation