Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Baclofen-D4 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco B0671ML

Catalog No. 01-001-054


Add to Cart

Description

Description

Baclofen, a derivative of gamma-aminobutyric acid (GABA), is a muscle relaxant used to treat spasticity. Marketed with brand names such as Kemstro, Lioresal or Gablofen, the drug has also shown effectiveness in the treatment of alcoholism. This certified solution standard is suitable for use as an internal standard in quantitative mass spectrometry-based testing applications in clinical toxicology, forensic analysis, pharmaceutical research or diagnostic testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
MFCD21363459
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C10H8D4ClNO2
UN 1230 - class 3 - PG 2 - Methanol, solution
217.69
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation