Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Benzodiazepine Multi-component Mixture-8 Solution, 250 μg/mL each in acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258405 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-405 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-405 Supplier MilliporeSigma Supelco Supplier No. B0331ML
Add to Cart
Add to Cart

A Certified Spiking Solution™ of routinely tested benzodiazepines alprazolam, clonazepam, diazepam, flunitrazepam, lorazepam, nitrazepam, oxazepam and temazepam.

This multi-component mix is suitable for use in preparation of linearity standards, calibrators, or controls as well as for other critical quantitative applications such as internal proficiency testing or for OEM use in preparation of reagents, assay development, and instrument IQ/OQ. Certified concentration values for each component provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
Product Line CERILLIANT
Physical Form Liquid
Concentration 250 μg/mL each component in Acetonitrile
Industry Type Forensics, Toxicology
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.