Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ beta-Lactoglobulin from bovine milk, certified reference material, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001550 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-550 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-550 Supplier MilliporeSigma Supelco Supplier No. L046100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

β-Lactoglobulin A & B is the major whey protein of cow and sheep's milk and is also present in many other mammalian species with an interesting exception being humans. This full-length protein CRM is suitable as starting material for use in calibrators or controls for a variety of LC-MS/MS testing applications including allergen testing, infant formula testing and dietary or nutritional testing applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 9045-23-2
MDL Number MFCD00130635
Packaging Ampule
Quantity 100 mg

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.