Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ BSTFA (with 1 TMCS) Kit, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111016625 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6625 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6625 Supplier MilliporeSigma Supelco Supplier No. B02310X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 25561-30-2
Product Line CERILLIANT
Density 0.969 g/mL (at 25°C)
Physical Form Neat
Linear Formula CF3C[=NSi(CH3)3 ]OSi(CH3)3
MDL Number MFCD00008269
Boiling Point 45°C to 50°C (14 mmHg)
Synonym N,O-Bis(trimethylsilyl)trifluoroacetamide; BSTFA
Quantity 10 x 1 mL
Formula Weight 257.40
Grade Analytical Standard
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.