Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Buprenorphine Solution, 100 μg/mL in methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258361 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-361 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-361 Supplier MilliporeSigma Supelco Supplier No. B9021ML
Add to Cart
Add to Cart

This Certified Spiking Solution™ standard is suitable for GC/MS or LC/MS applications in forensic analysis, clinical toxicology or pharmaceutical research.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
CAS 52485-79-7
Product Line CERILLIANT
Physical Form Liquid
Molecular Formula C29H41NO4
Concentration 100 μg/mL in Methanol
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 467.64
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.