Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Buspirone-D8 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001045 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-045 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-045 Supplier MilliporeSigma Supelco Supplier No. B0551ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A stable-labeled internal standard suitable for quantification of buspirone in urine, serum, and other biological matrices by LC-MS/MS or GC/MS for applications in clinical toxicology, urine drug testing, or forensic analysis. Buspirone, sold under the trade name Buspar™, is an antidepressant marketed for treatment of generalized anxiety disorder and clinical depression.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 204395-49-3
Linear Formula C21H23D8N5O2 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 430.01
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.