Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Buspirone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001044 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-044 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-044 Supplier MilliporeSigma Supelco Supplier No. B0541ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Buspirone, sold under the trade name Buspar™, is an antidepressant marketed for treatment of generalized anxiety disorder and clinical depression. Suitable uses for this Certified Spiking Solution™include as starting material for calibrators and controls in antidepressant testing methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 33386-08-2
Linear Formula C21H31N5O2 · HCl
MDL Number MFCD00078569
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 421.96
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.