Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Butyryl fentanyl-D5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001061 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-061 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-061 Supplier MilliporeSigma Supelco Supplier No. B0810.5ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Butyryl fentanyl is a designer drug and fentanyl analog that has been associated with numerous opioid overdose deaths. In May 2016, the U.S Drug Enforcement Administration (DEA) classified butyryl fentanyl as a schedule I controlled substance. A certified Snap-N-Shoot™ solution suitable for quantification of butyryl fentanyl by LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. This Certified Spiking Solution™ is supplied in a convenient US DEA-exempt solution format.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C23H25D5N2O
Concentration 100 μg/mL in methanol
Formula Weight 355.53
Packaging Ampule
Quantity 0.5 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.