Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cannabidivarin Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C1401ML

Catalog No. 01-001-099


 Restricted Item
Add to Cart

Description

Description

Cannabidivarin (CBDV) is a non-psychoactive cannabinoid found in Cannabis that reportedly has therapeutic efficacy in the treatment of pain, mood disorders, and inflammatory diseases. This certified Snap-N-Spike™ solution is suitable for cannabidivarin testing methods by GC/MS, LC/MS or HPLC for applications in clinical toxicology, testing of Cannabis potency or impurity profiling, pharmaceutical research, forensic analysis, and urine drug testing. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK's for Canadian customers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 24274-48-4
UN 1230 - class 3 - PG 2 - Methanol, solution
CBDV solution
Ampule
C19H26O2
1.0 mg/mL in methanol
286.41
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370-H412

H311

Recommended Storage : -70°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation