Learn More
CERILLIANT™ Cannabigerol-D3 (CBG-D3) Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco C2121ML
Catalog No. 01-001-144
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
67-56-1
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C21H29D3O2
319.50
1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation