Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cannabigerol Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001100 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-100 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-100 Supplier MilliporeSigma Supelco Supplier No. C1411ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A non-psychoactive cannabinoid found in Cannabis, Cannabigerol (CBG) reportedly has efficacy for medical conditions such as glaucoma and inflammatory bowel disease. This Certified Spiking Solution™ is suitable for cannabigerol testing methods by HPLC, GC/MS or LC-MS/MS for forensic analysis, clinical toxicology, urine drug testing, and Cannabis potency or impurity profiling testing applications. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK's for Canadian customers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 25654-31-3
Linear Formula C21H32O2
MDL Number MFCD00869599
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym CBG solution
Formula Weight 316.48
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.