Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Carbamazepine-10,11-epoxide Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001083 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-083 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-083 Supplier MilliporeSigma Supelco Supplier No. C1211ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Carbamazepine-10,11-epoxide is the primary, active metabolite in both urine and serum of carbamazepine, an anticonvulsant drug used in the treatment of epilepsy and bipolar disorder as well as trigeminal neuralgia. Potential toxicity of the epoxide metabolite requires regular therapeutic drug monitoring by LC or LC/MS for patients taking carbamazepine.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 36507-30-9
Linear Formula C15H12N2O2
MDL Number MFCD00467137
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 252.27
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.